missing translation for 'onlineSavingsMsg'
Learn More

o-Tolidine, 95%, pract

Catalog No. p-201036
Change view
Click to view available options
Quantity:
25 g
100 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC421120250 25 g
AC421121000 100 g
AC421122500 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC421120250 Supplier Thermo Scientific Chemicals Supplier No. 421120250
Only null left

CAS: 119-93-7 | C14H16N2 | 212.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator

Chemical Identifiers

CAS 119-93-7
MDL Number MFCD00014773
InChI Key NUIURNJTPRWVAP-UHFFFAOYSA-N
Synonym o-tolidine, 3,3'-dimethylbenzidine, orthotolidine, diaminoditolyl, 2-tolidine, diaminotolyl, bianisidine, 3,3'-tolidine, 3,3'-dimethylbiphenyl-4,4'-diamine, 4,4'-bi-o-toluidine
PubChem CID 8413
ChEBI CHEBI:34320
IUPAC Name 4-(4-amino-3-methylphenyl)-2-methylaniline
SMILES CC1=C(C=CC(=C1)C2=CC(=C(C=C2)N)C)N

Specifications

Melting Point 125.0°C to 132.0°C
Color Beige or Brown to Gray
Boiling Point 300.5°C
Flash Point 244°C
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Molecular Formula C14H16N2
Quantity 25 g
Beilstein 13, IV, 419
Merck Index 15, 9674
Molecular Weight (g/mol) 212.29
Formula Weight 212.29
Percent Purity 95%
Physical Form Powder
Chemical Name or Material o-Tolidine, (pract), 95%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May cause cancer.
Toxic to aquatic life with long lasting effects.
Harmful if swallowed.
Precautionary Statement GHS P Statement
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Avoid release to the environment.
EINECSNumber 204-358-0
RTECSNumber DD1225000
TSCA TSCA

RUO – Research Use Only