Learn More
o-Thiocresol, 98%
CAS: 137-06-4 | C7H8S | 124.20 g/mol
$76.44 - $789.10
Chemical Identifiers
| CAS | 137-06-4 |
|---|---|
| Molecular Formula | C7H8S |
| Molecular Weight (g/mol) | 124.20 |
| MDL Number | MFCD00004838 |
| InChI Key | LXUNZSDDXMPKLP-UHFFFAOYSA-N |
| Synonym | o-toluenethiol, o-thiocresol, 2-methylthiophenol, 2-thiocresol, 2-toluenethiol, o-tolyl mercaptan, o-mercaptotoluene, o-methylthiophenol, o-methylbenzenethiol, benzenethiol, 2-methyl |
| PubChem CID | 8712 |
| IUPAC Name | 2-methylbenzenethiol |
| SMILES | CC1=CC=CC=C1S |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAB2246006
|
Thermo Scientific Chemicals
B2246006 |
5 g |
Each for $76.44
|
|
|||||
|
AAB2246014
|
Thermo Scientific Chemicals
B2246014 |
25 g |
Each for $247.14
|
|
|||||
|
AAB2246022
|
Thermo Scientific Chemicals
B2246022 |
100 g |
Each for $789.10
|
|
|||||
Description
o-Thiocresol is used as a thiol for proteomics research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applicationso-Thiocresol is used as a thiol for proteomics research.
Solubility
Soluble in ether and ethanol. Insoluble in water.
Notes
Air Sensitive. Keep container tightly closed. Store away from oxidizing agents.
Chemical Identifiers
| 137-06-4 | |
| 124.20 | |
| LXUNZSDDXMPKLP-UHFFFAOYSA-N | |
| 8712 | |
| CC1=CC=CC=C1S |
| C7H8S | |
| MFCD00004838 | |
| o-toluenethiol, o-thiocresol, 2-methylthiophenol, 2-thiocresol, 2-toluenethiol, o-tolyl mercaptan, o-mercaptotoluene, o-methylthiophenol, o-methylbenzenethiol, benzenethiol, 2-methyl | |
| 2-methylbenzenethiol |
Specifications
| 137-06-4 | |
| 1.054 | |
| 63°C (145°F) | |
| 1.58 | |
| 5 g | |
| 774068 | |
| 14,9325 | |
| Soluble in ether and ethanol. Insoluble in water. | |
| CC1=CC=CC=C1S | |
| 124.20 | |
| 124.21 | |
| o-Thiocresol |
| 10°C to 12°C | |
| 195°C | |
| C7H8S | |
| MFCD00004838 | |
| UN2810 | |
| Air Sensitive | |
| o-toluenethiol, o-thiocresol, 2-methylthiophenol, 2-thiocresol, 2-toluenethiol, o-tolyl mercaptan, o-mercaptotoluene, o-methylthiophenol, o-methylbenzenethiol, benzenethiol, 2-methyl | |
| LXUNZSDDXMPKLP-UHFFFAOYSA-N | |
| 2-methylbenzenethiol | |
| 8712 | |
| 98% |
Safety and Handling
GHS H Statement
H331-H318-H302-H312-H315-H335
Toxic if inhaled.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
May cause respiratory irritation.
P210-P235-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P370+P378q-P501c
H227-H302+H312+H332-H315-H318-H335
DOTInformation : Hazard Class: 9; Packaging Group: III
EINECSNumber : 205-276-8
RTECSNumber : XT8750000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only