missing translation for 'onlineSavingsMsg'
Learn More

O-tert-Butyl-L-serine, 98%

CAS: 18822-58-7 | C7H15NO3 | 161.201 g/mol

$85.26 - $324.82

Chemical Identifiers

CAS 18822-58-7
Molecular Formula C7H15NO3
Molecular Weight (g/mol) 161.201
MDL Number MFCD00066088
InChI Key DDCPKNYKNWXULB-YFKPBYRVSA-N
Synonym h-ser tbu-oh, o-tert-butyl-l-serine, o-t-butyl-l-serine, s-2-amino-3-tert-butoxy propanoic acid, l-serine, o-1,1-dimethylethyl, 2s-2-amino-3-tert-butoxy propanoic acid, 2s-2-amino-3-2-methylpropan-2-yl oxy propanoic acid, ser tbu-oh, h-ser but-oh
PubChem CID 7004940
IUPAC Name (2S)-2-amino-3-[(2-methylpropan-2-yl)oxy]propanoic acid
SMILES CC(C)(C)OCC(C(=O)O)N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6381203
View Documents
Thermo Scientific Chemicals
H6381203
1 g
Each for $85.26
Only null left
 
AAH6381206
View Documents
Thermo Scientific Chemicals
H6381206
5 g
Each for $324.82
Only null left
 
Description

Description

O-tert-Butyl-L-serine tert-butyl ester hydrochloride is a protected L-serine used in peptide synthesis, e.g. synthesis of glycopeptides and lipopeptides and preparation of amino-phospholipids. O-tert-Butyl-L-serine (Ser(TBU-OH), a serine hydroxyl protected amino acid, is used in SPPS such as the synthesis of tailed cyclic RGD peptides to prevent glutarimide formation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
O-tert-Butyl-L-serine tert-butyl ester hydrochloride is a protected L-serine used in peptide synthesis, e.g. synthesis of glycopeptides and lipopeptides and preparation of amino-phospholipids. O-tert-Butyl-L-serine (Ser(TBU-OH), a serine hydroxyl protected amino acid, is used in SPPS such as the synthesis of tailed cyclic RGD peptides to prevent glutarimide formation.

Solubility
Soluble in water.

Notes
Light Sensitive.Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, light, heat.
Specifications

Chemical Identifiers

18822-58-7
161.201
DDCPKNYKNWXULB-YFKPBYRVSA-N
7004940
CC(C)(C)OCC(C(=O)O)N
C7H15NO3
MFCD00066088
h-ser tbu-oh, o-tert-butyl-l-serine, o-t-butyl-l-serine, s-2-amino-3-tert-butoxy propanoic acid, l-serine, o-1,1-dimethylethyl, 2s-2-amino-3-tert-butoxy propanoic acid, 2s-2-amino-3-2-methylpropan-2-yl oxy propanoic acid, ser tbu-oh, h-ser but-oh
(2S)-2-amino-3-[(2-methylpropan-2-yl)oxy]propanoic acid

Specifications

220°C (decomposition)
White
MFCD00066088
h-ser tbu-oh, o-tert-butyl-l-serine, o-t-butyl-l-serine, s-2-amino-3-tert-butoxy propanoic acid, l-serine, o-1,1-dimethylethyl, 2s-2-amino-3-tert-butoxy propanoic acid, 2s-2-amino-3-2-methylpropan-2-yl oxy propanoic acid, ser tbu-oh, h-ser but-oh
DDCPKNYKNWXULB-YFKPBYRVSA-N
(2S)-2-amino-3-[(2-methylpropan-2-yl)oxy]propanoic acid
161.201
161.2
Powder
18822-58-7
C7H15NO3
2500118
Soluble in water.
CC(C)(C)OCC(C(=O)O)N
1 g
7004940
98%
O-tert-Butyl-L-serine
Videos
Safety and Handling

Safety and Handling

TSCA : No

storageNote1 : 2° to 8°C

Recommended Storage : Keep cold

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only