accessibility menu, dialog, popup

UserName

CAS RN 18822-58-7

IUPAC Name: 2-amino-3-(tert-butoxy)propanoic acid
Synonyms: 2-amino-3-(tert-butoxy)propanoic acid
Molecular Weight (g/mol): 161.2
Molecular Formula: C7H15NO3
InChi Key: DDCPKNYKNWXULB-UHFFFAOYNA-N
SMILES: CC(C)(C)OCC(N)C(O)=O
Molecular Weight (g/mol) 
  • (2)
  • (4)
Percent Purity 
  • (2)
  • (2)
  • (2)
Quantity 
  • (3)
  • (3)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (4)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (3)
  • (3)
  • (4)