Learn More
O-tert-Butyl-L-serine, 98%
CAS: 18822-58-7 | C7H15NO3 | 161.201 g/mol
Supplier: Thermo Scientific Chemicals H6381203
Description
O-tert-Butyl-L-serine tert-butyl ester hydrochloride is a protected L-serine used in peptide synthesis, e.g. synthesis of glycopeptides and lipopeptides and preparation of amino-phospholipids. O-tert-Butyl-L-serine (Ser(TBU-OH), a serine hydroxyl protected amino acid, is used in SPPS such as the synthesis of tailed cyclic RGD peptides to prevent glutarimide formation.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsO-tert-Butyl-L-serine tert-butyl ester hydrochloride is a protected L-serine used in peptide synthesis, e.g. synthesis of glycopeptides and lipopeptides and preparation of amino-phospholipids. O-tert-Butyl-L-serine (Ser(TBU-OH), a serine hydroxyl protected amino acid, is used in SPPS such as the synthesis of tailed cyclic RGD peptides to prevent glutarimide formation.
Solubility
Soluble in water.
Notes
Light Sensitive.Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, light, heat.
Specifications
| 220°C (decomposition) | |
| 18822-58-7 | |
| C7H15NO3 | |
| 2500118 | |
| Soluble in water. | |
| CC(C)(C)OCC(C(=O)O)N | |
| 1 g | |
| 7004940 | |
| 98% |
| O-tert-Butyl-L-serine | |
| White | |
| MFCD00066088 | |
| h-ser tbu-oh, o-tert-butyl-l-serine, o-t-butyl-l-serine, s-2-amino-3-tert-butoxy propanoic acid, l-serine, o-1,1-dimethylethyl, 2s-2-amino-3-tert-butoxy propanoic acid, 2s-2-amino-3-2-methylpropan-2-yl oxy propanoic acid, ser tbu-oh, h-ser but-oh | |
| DDCPKNYKNWXULB-YFKPBYRVSA-N | |
| (2S)-2-amino-3-[(2-methylpropan-2-yl)oxy]propanoic acid | |
| 161.201 | |
| 161.2 | |
| Powder |
Chemical Identifiers
| 18822-58-7 | |
| 161.201 | |
| DDCPKNYKNWXULB-YFKPBYRVSA-N | |
| 7004940 | |
| CC(C)(C)OCC(C(=O)O)N |
| C7H15NO3 | |
| MFCD00066088 | |
| h-ser tbu-oh, o-tert-butyl-l-serine, o-t-butyl-l-serine, s-2-amino-3-tert-butoxy propanoic acid, l-serine, o-1,1-dimethylethyl, 2s-2-amino-3-tert-butoxy propanoic acid, 2s-2-amino-3-2-methylpropan-2-yl oxy propanoic acid, ser tbu-oh, h-ser but-oh | |
| (2S)-2-amino-3-[(2-methylpropan-2-yl)oxy]propanoic acid |
Safety and Handling
TSCA : No
storageNote1 : 2° to 8°C
Recommended Storage : Keep cold
RUO – Research Use Only