missing translation for 'onlineSavingsMsg'
Learn More
Learn More
O-tert-Butyl-L-serine, 97%, Thermo Scientific™
Chemical Identifiers
| CAS | 18822-58-7 |
|---|---|
| Molecular Formula | C7H15NO3 |
| Molecular Weight (g/mol) | 161.2 |
| InChI Key | DDCPKNYKNWXULB-YFKPBYRVSA-N |
| Synonym | h-ser tbu-oh, o-tert-butyl-l-serine, o-t-butyl-l-serine, s-2-amino-3-tert-butoxy propanoic acid, l-serine, o-1,1-dimethylethyl, 2s-2-amino-3-tert-butoxy propanoic acid, 2s-2-amino-3-2-methylpropan-2-yl oxy propanoic acid, ser tbu-oh, h-ser but-oh |
| PubChem CID | 7004940 |
| IUPAC Name | (2S)-2-amino-3-[(2-methylpropan-2-yl)oxy]propanoic acid |
| SMILES | CC(C)(C)OCC(C(=O)O)N |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AC434880010
|
Thermo Scientific Chemicals
434880010 |
1g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AC434880050
|
Thermo Scientific Chemicals
434880050 |
5g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 18822-58-7 | |
| 161.2 | |
| h-ser tbu-oh, o-tert-butyl-l-serine, o-t-butyl-l-serine, s-2-amino-3-tert-butoxy propanoic acid, l-serine, o-1,1-dimethylethyl, 2s-2-amino-3-tert-butoxy propanoic acid, 2s-2-amino-3-2-methylpropan-2-yl oxy propanoic acid, ser tbu-oh, h-ser but-oh | |
| (2S)-2-amino-3-[(2-methylpropan-2-yl)oxy]propanoic acid |
| C7H15NO3 | |
| DDCPKNYKNWXULB-YFKPBYRVSA-N | |
| 7004940 | |
| CC(C)(C)OCC(C(=O)O)N |
Specifications
| 18822-58-7 | |
| 96% min. (HPLC) | |
| C7H15NO3 | |
| − 15.50 | |
| DDCPKNYKNWXULB-YFKPBYRVSA-N | |
| (2S)-2-amino-3-[(2-methylpropan-2-yl)oxy]propanoic acid | |
| 7004940 | |
| 97% |
| Authentic | |
| Glass Bottle | |
| 1g | |
| h-ser tbu-oh, o-tert-butyl-l-serine, o-t-butyl-l-serine, s-2-amino-3-tert-butoxy propanoic acid, l-serine, o-1,1-dimethylethyl, 2s-2-amino-3-tert-butoxy propanoic acid, 2s-2-amino-3-2-methylpropan-2-yl oxy propanoic acid, ser tbu-oh, h-ser but-oh | |
| CC(C)(C)OCC(C(=O)O)N | |
| 161.2 | |
| 161.2 | |
| O-tert-Butyl-L-serine |
RUO â Research Use Only