missing translation for 'onlineSavingsMsg'
Learn More

O-tert-Butyl-L-serine, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 434880010

 View more versions of this product

Catalog No. AC434880010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

O-tert-Butyl-L-serine
Authentic
Glass Bottle
1g
h-ser tbu-oh, o-tert-butyl-l-serine, o-t-butyl-l-serine, s-2-amino-3-tert-butoxy propanoic acid, l-serine, o-1,1-dimethylethyl, 2s-2-amino-3-tert-butoxy propanoic acid, 2s-2-amino-3-2-methylpropan-2-yl oxy propanoic acid, ser tbu-oh, h-ser but-oh
CC(C)(C)OCC(C(=O)O)N
161.2
161.2
18822-58-7
96% min. (HPLC)
C7H15NO3
− 15.50
DDCPKNYKNWXULB-YFKPBYRVSA-N
(2S)-2-amino-3-[(2-methylpropan-2-yl)oxy]propanoic acid
7004940
97%

Chemical Identifiers

18822-58-7
161.2
h-ser tbu-oh, o-tert-butyl-l-serine, o-t-butyl-l-serine, s-2-amino-3-tert-butoxy propanoic acid, l-serine, o-1,1-dimethylethyl, 2s-2-amino-3-tert-butoxy propanoic acid, 2s-2-amino-3-2-methylpropan-2-yl oxy propanoic acid, ser tbu-oh, h-ser but-oh
(2S)-2-amino-3-[(2-methylpropan-2-yl)oxy]propanoic acid
C7H15NO3
DDCPKNYKNWXULB-YFKPBYRVSA-N
7004940
CC(C)(C)OCC(C(=O)O)N
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only