missing translation for 'onlineSavingsMsg'
Learn More

o-Phthalaldehyde, Spectrum™ Chemical

Catalog No. 18612082
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
Packaging:
Amber Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18612082 1 g Amber Glass Bottle
18612085 5 g Amber Glass Bottle
18612084 25 g Amber Glass Bottle
18612083 100 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18612082 Supplier Spectrum Chemical Mfg Cor Supplier No. P11391GM
Get Up to $500 in Chemicals with Purchase
View all promotions

Chemical Identifiers

CAS 643-79-8
Molecular Formula C8H6O2
Molecular Weight (g/mol) 134.13
InChI Key ZWLUXSQADUDCSB-UHFFFAOYSA-N
IUPAC Name benzene-1,2-dicarbaldehyde
SMILES O=CC1=CC=CC=C1C=O

Specifications

CAS Max % 1
Melting Point 53° to 58°C
Boiling Point 83°C
Packaging Amber Glass Bottle
Quantity 1 g
Grade Reagent
Physical Form Crystals or Flakes

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.