accessibility menu, dialog, popup

UserName

OO

CAS RN 643-79-8

IUPAC Name: benzene-1,2-dicarbaldehyde
Synonyms: phthalaldehyde ortho phthalic aldehyde o-phthaldialdehyde ortho-phthalic aldehyde aldehyde, ortho-phthalic phtalaldehydes o-phthalaldehyde phthalyldicarboxaldehyde phtharal o phthalaldehyde
Molecular Weight (g/mol): 134.13
Molecular Formula: C8H6O2
InChi Key: ZWLUXSQADUDCSB-UHFFFAOYSA-N
SMILES: O=CC1=CC=CC=C1C=O
Quantity 
  • (1)
  • (1)
  • (3)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
Percent Purity 
  • (3)
  • (3)
  • (4)
Boiling Point 
  • (4)
  • (4)
Physical Form 
  • (4)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (11)
  • (4)
  • (4)
  • (3)
  • (7)
  • (1)
  • (1)
  • (3)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (3)
  • (3)
  • (4)
  • (4)
  • (4)
  • (4)
  • (4)
  • (4)