missing translation for 'onlineSavingsMsg'
Learn More

O-Phosphorylethanolamine 98.0+%, TCI America™

Supplier:  TCI America P027925G

 View more versions of this product

Catalog No. P027925G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

O-Phosphorylethanolamine
235°C
C2H8NO4P
25 g
o-phosphoethanolamine, o-phosphorylethanolamine, phosphoethanolamine, o-phosphocolamine, colamine phosphate, phosphorylethanolamine, ethanolamine phosphate, phosphonoethanolamine, ethanolamine o-phosphate, mono 2-aminoethyl phosphate
C(COP(=O)(O)O)N
141.063
CHEBI:17553
≥98.0% (N,T)
1071-23-4
White-Yellow
MFCD00008178
1759
SUHOOTKUPISOBE-UHFFFAOYSA-N
2-aminoethyl dihydrogen phosphate
1015
141.06
Crystalline Powder

Chemical Identifiers

1071-23-4
141.063
SUHOOTKUPISOBE-UHFFFAOYSA-N
1015
2-aminoethyl dihydrogen phosphate
C2H8NO4P
MFCD00008178
o-phosphoethanolamine, o-phosphorylethanolamine, phosphoethanolamine, o-phosphocolamine, colamine phosphate, phosphorylethanolamine, ethanolamine phosphate, phosphonoethanolamine, ethanolamine o-phosphate, mono 2-aminoethyl phosphate
CHEBI:17553
C(COP(=O)(O)O)N
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-2917

RTECSNumber : KJ5830000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions