missing translation for 'onlineSavingsMsg'
Learn More

O-Phosphorylethanolamine 98.0+%, TCI America™

Catalog No. p-7134530
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P027925G 25 g
P0279100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P027925G Supplier TCI America Supplier No. P027925G
Only null left

Chemical Identifiers

CAS 1071-23-4
Molecular Formula C2H8NO4P
Molecular Weight (g/mol) 141.063
MDL Number MFCD00008178
InChI Key SUHOOTKUPISOBE-UHFFFAOYSA-N
Synonym o-phosphoethanolamine, o-phosphorylethanolamine, phosphoethanolamine, o-phosphocolamine, colamine phosphate, phosphorylethanolamine, ethanolamine phosphate, phosphonoethanolamine, ethanolamine o-phosphate, mono 2-aminoethyl phosphate
PubChem CID 1015
ChEBI CHEBI:17553
IUPAC Name 2-aminoethyl dihydrogen phosphate
SMILES C(COP(=O)(O)O)N

Specifications

Melting Point 235°C
Color White-Yellow
Quantity 25 g
UN Number 1759
Formula Weight 141.06
Percent Purity ≥98.0% (N,T)
Physical Form Crystalline Powder
Chemical Name or Material O-Phosphorylethanolamine
EINECSNumber (2)-2917
RTECSNumber KJ5830000
TSCA Yes