Learn More
o-Phenylenediacetonitrile, 99+%
CAS: 613-73-0 | C10H8N2 | 156.19 g/mol
$43.38 - $43.38
Chemical Identifiers
| CAS | 613-73-0 |
|---|---|
| Molecular Formula | C10H8N2 |
| Molecular Weight (g/mol) | 156.19 |
| MDL Number | MFCD00001905 |
| InChI Key | FWPFXBANOKKNBR-UHFFFAOYSA-N |
| Synonym | 1,2-phenylenediacetonitrile, o-phenylenediacetonitrile, 1,2-bis cyanomethyl benzene, o-xylylene dicyanide, 2-2-cyanomethyl phenyl acetonitrile, o-benzenediacetonitrile, 2,2'-1,2-phenylene diacetonitrile, o-bis cyanomethyl benzene, 1,2-benzenediacetonitrile, o-benzendiacetonitrile |
| PubChem CID | 69180 |
| IUPAC Name | 2-[2-(cyanomethyl)phenyl]acetonitrile |
| SMILES | N#CCC1=CC=CC=C1CC#N |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC130520100
|
Thermo Scientific Chemicals
130520100 |
10 g | Glass bottle |
Each for $43.38
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 613-73-0 | |
| 156.19 | |
| FWPFXBANOKKNBR-UHFFFAOYSA-N | |
| 69180 | |
| N#CCC1=CC=CC=C1CC#N |
| C10H8N2 | |
| MFCD00001905 | |
| 1,2-phenylenediacetonitrile, o-phenylenediacetonitrile, 1,2-bis cyanomethyl benzene, o-xylylene dicyanide, 2-2-cyanomethyl phenyl acetonitrile, o-benzenediacetonitrile, 2,2'-1,2-phenylene diacetonitrile, o-bis cyanomethyl benzene, 1,2-benzenediacetonitrile, o-benzendiacetonitrile | |
| 2-[2-(cyanomethyl)phenyl]acetonitrile |
Specifications
| 613-73-0 | |
| 100.0 | |
| Authentic | |
| Glass bottle | |
| C6H4(CH2CN)2 | |
| 10 g | |
| 1,2-phenylenediacetonitrile, o-phenylenediacetonitrile, 1,2-bis cyanomethyl benzene, o-xylylene dicyanide, 2-2-cyanomethyl phenyl acetonitrile, o-benzenediacetonitrile, 2,2'-1,2-phenylene diacetonitrile, o-bis cyanomethyl benzene, 1,2-benzenediacetonitrile, o-benzendiacetonitrile | |
| FWPFXBANOKKNBR-UHFFFAOYSA-N | |
| 2-[2-(cyanomethyl)phenyl]acetonitrile | |
| 69180 | |
| 99+% | |
| o-Phenylenediacetonitrile, 99+% |
| 99.0 | |
| 58°C to 61°C | |
| 99% min. (GC) | |
| C10H8N2 | |
| MFCD00001905 | |
| 09, 874 | |
| Solubility in water: insoluble | |
| N#CCC1=CC=CC=C1CC#N | |
| 156.19 | |
| 156.19 | |
| Crystalline Powder, Crystals or Chunks |
Safety and Handling
GHS H Statement:
Harmful if inhaled.
Harmful in contact with skin.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
EINECSNumber : 210-351-3
RUO – Research Use Only