accessibility menu, dialog, popup

UserName

NN

CAS RN 613-73-0

IUPAC Name: 2-[2-(cyanomethyl)phenyl]acetonitrile
Synonyms: 2-[2-(cyanomethyl)phenyl]acetonitrile
Molecular Weight (g/mol): 156.19
Molecular Formula: C10H8N2
InChi Key: FWPFXBANOKKNBR-UHFFFAOYSA-N
SMILES: N#CCC1=CC=CC=C1CC#N
Percent Purity 
  • (2)
  • (3)
  • (2)
Quantity 
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)