missing translation for 'onlineSavingsMsg'
Learn More

o-Phenanthroline Solution, Certified, 0.25% (w/v), LabChem™

Supplier:  LabChem LC181601

Catalog No. LC181601

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

o-Phenanthroline Solution
5144-89-8 , 7732-18-5
C12H10N2O
MFCD00149973
1,10-phenanthroline hydrate, 1,10-phenanthroline monohydrate, o-phenanthroline monohydrate, unii-ksx215x00e, 1,10-fenantrolina, 4,5-phenanthroline monohydrate, 1,10-phenanthroline, monohydrate, 1,10-fenanthrolin, 1,10-phenanthrolineo-phenanthroline, 1, 10-phenanthroline monohydrate
PPQJCISYYXZCAE-UHFFFAOYSA-N
1,10-phenanthroline hydrate
21226
0.25% (w/v)
Passes Test
1g/mL
Carbon monoxide; Carbon dioxide
Liquid
Colorless
99.75,0.25
C12H8N2·H2O
1g/mL
Soluble in water
O.C1=CN=C2C(C=CC3=CC=CN=C23)=C1
198.23
198.22
Certified
Poly Bottle
Passes Test
500 mL

Chemical Identifiers

5144-89-8
198.23
PPQJCISYYXZCAE-UHFFFAOYSA-N
21226
O.C1=CN=C2C(C=CC3=CC=CN=C23)=C1
C12H10N2O
MFCD00149973
1,10-phenanthroline hydrate, 1,10-phenanthroline monohydrate, o-phenanthroline monohydrate, unii-ksx215x00e, 1,10-fenantrolina, 4,5-phenanthroline monohydrate, 1,10-phenanthroline, monohydrate, 1,10-fenanthrolin, 1,10-phenanthrolineo-phenanthroline, 1, 10-phenanthroline monohydrate
1,10-phenanthroline hydrate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful to aquatic life with long lasting effects.

GHS P Statement
Avoid release to the environment.
Dispose of contents/container to comply with local, state and federal regulations.

Recommended Storage : Room Temperature

SDS
Documents

Documents

Promotions

Promotions