missing translation for 'onlineSavingsMsg'
Learn More

o-Nitrophenol, 99%, Spectrum™ Chemical

Catalog No. 18612079
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
100 g
250 kg
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18612079 25 g Amber Glass Bottle
18612078 100 g Amber Glass Bottle
18612080 250 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 missing translation for 'options'
Catalog No. 18612079 missing translation for 'mfr' Spectrum Chemical Mfg Cor missing translation for 'supplierNo' N109825GM
Get Up to $500 in Chemicals with Purchase
View all promotions

Chemical Identifiers

CAS 88-75-5

Specifications

CAS Max % 1
Boiling Point 215°C
Melting Range 43° to 51°C
Packaging Amber Glass Bottle
Quantity 25 g
Percent Purity 0.99
Grade Reagent
Physical Form Crystalline Powder or Lumps

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.