accessibility menu, dialog, popup

UserName

OHNOO

CAS RN 88-75-5

IUPAC Name: 2-nitrophenol
Synonyms: o-nitrophenol
Molecular Weight (g/mol): 139.11
Molecular Formula: C6H5NO3
InChi Key: IQUPABOKLQSFBK-UHFFFAOYSA-N
SMILES: OC1=CC=CC=C1[N+]([O-])=O
Boiling Point 
  • (2)
  • (4)
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)
  • (2)
  • (4)
Quantity 
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (3)
  • (3)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (9)
  • (3)
  • (3)
  • (3)
  • (6)
  • (2)
  • (4)
  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (4)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (3)
  • (4)
  • (3)