missing translation for 'onlineSavingsMsg'
Learn More

o-Nitrophenol, 99%, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor N1098100GM

 View more versions of this product

Catalog No. 18-612-078


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

88-75-5
215°C
Amber Glass Bottle
0.99
Crystalline Powder or Lumps
1
43° to 51°C
100 g
Reagent

Chemical Identifiers

88-75-5
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.