missing translation for 'onlineSavingsMsg'
Learn More
Learn More
O-Methyl-L-serine hydrochloride, 95%
CAS: 336100-47-1 | C4H10ClNO3 | 155.58 g/mol
Supplier: Thermo Scientific Chemicals H5256203
| Quantity | 1 g |
|---|
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 336100-47-1 | |
| 155.58 | |
| NLDVBPFPOCWZCE-UHFFFAOYNA-N | |
| 16038806 | |
| Cl.COCC(N)C(O)=O |
| C4H10ClNO3 | |
| MFCD03840366 | |
| s-2-amino-3-methoxypropanoic acid hydrochloride, s-2-amino-3-methoxy-propionic acid hydrochloride, s-2-amino-3-methoxypropionic acid hydrochloride, o-methyl-l-serine hydrochloride, 2s-2-amino-3-methoxypropanoic acid hydrochloride, l-serine, o-methyl-, hydrochloride 1:1, l-o-methylserine hydrochloride, l-serine, o-methyl-, hydrochloride, s-2-amino-3-methoxy-propionicacidhydrochloride, s-2-amino-3-methoxypropanoic acid hcl | |
| 2-amino-3-methoxypropanoic acid hydrochloride |
Specifications
| 182°C to 183°C | |
| 336100-47-1 | |
| MFCD03840366 | |
| NLDVBPFPOCWZCE-UHFFFAOYNA-N | |
| 2-amino-3-methoxypropanoic acid hydrochloride | |
| 155.58 | |
| 155.58 |
| O-Methyl-L-serine hydrochloride | |
| C4H10ClNO3 | |
| s-2-amino-3-methoxypropanoic acid hydrochloride, s-2-amino-3-methoxy-propionic acid hydrochloride, s-2-amino-3-methoxypropionic acid hydrochloride, o-methyl-l-serine hydrochloride, 2s-2-amino-3-methoxypropanoic acid hydrochloride, l-serine, o-methyl-, hydrochloride 1:1, l-o-methylserine hydrochloride, l-serine, o-methyl-, hydrochloride, s-2-amino-3-methoxy-propionicacidhydrochloride, s-2-amino-3-methoxypropanoic acid hcl | |
| Cl.COCC(N)C(O)=O | |
| 1 g | |
| 16038806 | |
| 97% |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures