CAS RN 336100-47-1
IUPAC Name:
2-amino-3-methoxypropanoic acid hydrochloride
Synonyms:
DL-serine, O-methyl- hydrochloride
Molecular Weight (g/mol):
155.58
Molecular Formula:
C4H10ClNO3
InChi Key:
NLDVBPFPOCWZCE-UHFFFAOYNA-N
SMILES:
Cl.COCC(N)C(O)=O
1
–
1
of
1
results
1
Promotions Available
O-Methyl-L-serine hydrochloride, 95%
O-Methyl-L-serine hydrochloride, 95%
CAS: 336100-47-1 Molecular Formula: C4H10ClNO3 Molecular Weight (g/mol): 155.58 MDL Number: MFCD03840366 InChI Key: NLDVBPFPOCWZCE-UHFFFAOYNA-N Synonym: s-2-amino-3-methoxypropanoic acid hydrochloride,s-2-amino-3-methoxy-propionic acid hydrochloride,s-2-amino-3-methoxypropionic acid hydrochloride,o-methyl-l-serine hydrochloride,2s-2-amino-3-methoxypropanoic acid hydrochloride,l-serine, o-methyl-, hydrochloride 1:1,l-o-methylserine hydrochloride,l-serine, o-methyl-, hydrochloride,s-2-amino-3-methoxy-propionicacidhydrochloride,s-2-amino-3-methoxypropanoic acid hcl PubChem CID: 16038806 IUPAC Name: 2-amino-3-methoxypropanoic acid hydrochloride SMILES: Cl.COCC(N)C(O)=O