missing translation for 'onlineSavingsMsg'
Learn More

O-Methyl-L-serine hydrochloride, 95%

Catalog No. p-7034862
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH52562MD 250 mg
AAH5256203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH52562MD Supplier Thermo Scientific Chemicals Supplier No. H52562MD
Only null left

CAS: 336100-47-1 | C4H10ClNO3 | 155.58 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 336100-47-1
Molecular Formula C4H10ClNO3
Molecular Weight (g/mol) 155.58
MDL Number MFCD03840366
InChI Key NLDVBPFPOCWZCE-UHFFFAOYNA-N
Synonym s-2-amino-3-methoxypropanoic acid hydrochloride, s-2-amino-3-methoxy-propionic acid hydrochloride, s-2-amino-3-methoxypropionic acid hydrochloride, o-methyl-l-serine hydrochloride, 2s-2-amino-3-methoxypropanoic acid hydrochloride, l-serine, o-methyl-, hydrochloride 1:1, l-o-methylserine hydrochloride, l-serine, o-methyl-, hydrochloride, s-2-amino-3-methoxy-propionicacidhydrochloride, s-2-amino-3-methoxypropanoic acid hcl
PubChem CID 16038806
IUPAC Name 2-amino-3-methoxypropanoic acid hydrochloride
SMILES Cl.COCC(N)C(O)=O

Specifications

Melting Point 182°C to 183°C
Quantity 250 mg
Formula Weight 155.58
Percent Purity 97%
Chemical Name or Material O-Methyl-L-serine hydrochloride
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only