missing translation for 'onlineSavingsMsg'
Learn More

o-Dianisidine 95.0+%, TCI America™

Supplier:  TCI America D38645G

 View more versions of this product

Catalog No. D38645G

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

o-Dianisidine [for Biochemical Research]
138°C
MFCD00008372
2811
JRBJSXQPQWSCCF-UHFFFAOYSA-N
4-(4-amino-3-methoxyphenyl)-2-methoxyaniline
8411
244.29
Crystalline Powder
119-90-4
C14H16N2O2
5 g
3,3'-dimethoxybenzidine, o-dianisidine, dianisidine, cellitazol b, neutrosel navy bn, diacel navy dc, disperse black 6, blue base nb, blue bn base, blue base irga b
COC1=C(C=CC(=C1)C2=CC(=C(C=C2)N)OC)N
244.294
CHEBI:82321
≥95.0% (T)

Chemical Identifiers

119-90-4
244.294
JRBJSXQPQWSCCF-UHFFFAOYSA-N
8411
4-(4-amino-3-methoxyphenyl)-2-methoxyaniline
C14H16N2O2
MFCD00008372
3,3'-dimethoxybenzidine, o-dianisidine, dianisidine, cellitazol b, neutrosel navy bn, diacel navy dc, disperse black 6, blue base nb, blue bn base, blue base irga b
CHEBI:82321
COC1=C(C=CC(=C1)C2=CC(=C(C=C2)N)OC)N
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (5)-2328

RTECSNumber : DD0875000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions