accessibility menu, dialog, popup

UserName

H3COH2NNH2OH3C

CAS RN 119-90-4

IUPAC Name: 3,3'-dimethoxy-[1,1'-biphenyl]-4,4'-diamine
Synonyms: dianisidine
Molecular Weight (g/mol): 244.29
Molecular Formula: C14H16N2O2
InChi Key: JRBJSXQPQWSCCF-UHFFFAOYSA-N
SMILES: COC1=CC(=CC=C1N)C1=CC=C(N)C(OC)=C1
Molecular Weight (g/mol) 
  • (2)
  • (7)
Percent Purity 
  • (2)
  • (1)
  • (3)
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (2)
  • (3)
  • (2)
Physical Form 
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (4)
  • (4)
  • (2)
  • (7)
  • (2)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (2)
  • (5)