missing translation for 'onlineSavingsMsg'
Learn More

o-Dianisidine 95.0+%, TCI America™

Catalog No. D38641G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D38641G 1 g
D38645G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D38641G Supplier TCI America Supplier No. D38641G

Chemical Identifiers

CAS 119-90-4
Molecular Formula C14H16N2O2
Molecular Weight (g/mol) 244.294
MDL Number MFCD00008372
InChI Key JRBJSXQPQWSCCF-UHFFFAOYSA-N
Synonym 3,3'-dimethoxybenzidine, o-dianisidine, dianisidine, cellitazol b, neutrosel navy bn, diacel navy dc, disperse black 6, blue base nb, blue bn base, blue base irga b
PubChem CID 8411
ChEBI CHEBI:82321
IUPAC Name 4-(4-amino-3-methoxyphenyl)-2-methoxyaniline
SMILES COC1=C(C=CC(=C1)C2=CC(=C(C=C2)N)OC)N

Specifications

Melting Point 138°C
Quantity 1 g
UN Number 2811
Formula Weight 244.29
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material o-Dianisidine [for Biochemical Research]
EINECSNumber (5)-2328
RTECSNumber DD0875000
TSCA Yes