missing translation for 'onlineSavingsMsg'
Learn More

o-Dianisidine dihydrochloride, 98%

Catalog No. p-7023529
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1717503 1 g
AAA1717506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1717503 Supplier Thermo Scientific Chemicals Supplier No. A1717503

CAS: 20325-40-0 | C14H18Cl2N2O2 | 317.21 g/mol

It is suitable for use in ELISA procedures. In neurohistochemistry o-Dianisidine dihydrochloride has demonstrated perikaryal labeling as well as retrograde and anterograde transport to the primary olfactory cortex. It has also been used for a paper based-micro fluidic device to semi-quantitatively measure lactate, uric acid, and glucose. 3,3'-Dimethoxybenzidine is used principally as an intermediate in the production of commercial bisazobiphenyl dyes for coloring textiles, paper, plastic, rubber, and leather.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is suitable for use in ELISA procedures. In neurohistochemistry o-Dianisidine dihydrochloride has demonstrated perikaryal labeling as well as retrograde and anterograde transport to the primary olfactory cortex. It has also been used for a paper based-micro fluidic device to semi-quantitatively measure lactate, uric acid, and glucose. 3,3′-Dimethoxybenzidine is used principally as an intermediate in the production of commercial bisazobiphenyl dyes for coloring textiles, paper, plastic, rubber, and leather.

Solubility
Soluble in water (50 mg/ml). It dissolves in ethyl alcohol ,acid and in organic solvent.

Notes
Store away from oxidizing agents. Keep the container tightly sealed and store in cool, dry condition in well ventilated place.

Chemical Identifiers

CAS 20325-40-0
Molecular Formula C14H18Cl2N2O2
Molecular Weight (g/mol) 317.21
MDL Number MFCD00012488
InChI Key UXTIAFYTYOEQHV-UHFFFAOYSA-N
Synonym 3,3'-dimethoxybenzidine dihydrochloride, o-dianisidine dihydrochloride, 3,3'-dimethoxy-1,1'-biphenyl-4,4'-diamine dihydrochloride, unii-qv96qa6ukk, dianisidine dihydrochloride, ccris 998, o-dianisidine hcl, qv96qa6ukk, c.i. disperse black 6 dihydrochloride
PubChem CID 62311
IUPAC Name 4-(4-amino-3-methoxyphenyl)-2-methoxyaniline;dihydrochloride
SMILES COC1=C(C=CC(=C1)C2=CC(=C(C=C2)N)OC)N.Cl.Cl

Specifications

Melting Point 268°C (decomposition)
Assay Percent Range 99%
Quantity 1 g
Beilstein 3917996
Solubility Information Soluble in water (50mg/ml). It dissolves in ethyl alcohol ,acid and in organic solvent.
Formula Weight 317.21
Percent Purity 99%
Chemical Name or Material o-Dianisidine dihydrochloride
Hazard Category H302-H314-H335-H350
Hazard Statement GHS H Statement
H350-H302
Precautionary Statement P201-P202-P260-P264b-P270-P271-P281-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
EINECSNumber 243-737-5
RTECSNumber DD1050000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only