accessibility menu, dialog, popup

UserName

HHClClH3COH2NNH2OH3C

CAS RN 20325-40-0

IUPAC Name: dihydrogen 3,3'-dimethoxy-[1,1'-biphenyl]-4,4'-diamine dichloride
Synonyms: dihydrogen dianisidine dichloride
Molecular Weight (g/mol): 317.21
Molecular Formula: C14H18Cl2N2O2
InChi Key: UXTIAFYTYOEQHV-UHFFFAOYSA-N
SMILES: [H+].[H+].[Cl-].[Cl-].COC1=CC(=CC=C1N)C1=CC=C(N)C(OC)=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results