missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals o-Cresolphthalein Complexone, pure
CAS: 2411-89-4 | C32H32N2O12 | 636.61 g/mol
Supplier: Thermo Scientific Chemicals 182440250
| Quantity | 25 g |
|---|---|
| Packaging | Glass bottle |
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
IndicatorChemical Identifiers
| 2411-89-4 | |
| 636.61 | |
| IYZPEGVSBUNMBE-UHFFFAOYSA-N | |
| 75485 | |
| CC1=CC(=CC(CN(CC(O)=O)CC(O)=O)=C1O)C1(OC(=O)C2=CC=CC=C12)C1=CC(C)=C(O)C(CN(CC(O)=O)CC(O)=O)=C1 |
| C32H32N2O12 | |
| MFCD00005911 | |
| o-Cresolphthalexon, Phthalein Complexon | |
| 2-[({5-[1-(3-{[bis(carboxymethyl)amino]methyl}-4-hydroxy-5-methylphenyl)-3-oxo-1,3-dihydro-2-benzofuran-1-yl]-2-hydroxy-3-methylphenyl}methyl)(carboxymethyl)amino]acetic acid |
Specifications
| o-Cresolphthalein Complexone | |
| Pure | |
| C32H32N2O12 | |
| 15, 2566 | |
| Solubility in water: slightly soluble. Other solubilities: readily soluble in aqueous ammonia, readily soluble in organic solvents | |
| CC1=CC(=CC(CN(CC(O)=O)CC(O)=O)=C1O)C1(OC(=O)C2=CC=CC=C12)C1=CC(C)=C(O)C(CN(CC(O)=O)CC(O)=O)=C1 | |
| 2-[({5-[1-(3-{[bis(carboxymethyl)amino]methyl}-4-hydroxy-5-methylphenyl)-3-oxo-1,3-dihydro-2-benzofuran-1-yl]-2-hydroxy-3-methylphenyl}methyl)(carboxymethyl)amino]acetic acid | |
| 75485 | |
| 636.6 | |
| Pure | |
| As indicator for complexometry: Passes Test | |
| Brown to White | |
| Powder |
| 181.0°C | |
| 2411-89-4 | |
| MFCD00005911 | |
| o-Cresolphthalexon, Phthalein Complexon | |
| IYZPEGVSBUNMBE-UHFFFAOYSA-N | |
| Authentic | |
| 636.61 | |
| 573 to 579nm (in 0.1N NaOH) | |
| 10% max. (1 g, 105°C) | |
| Glass bottle | |
| Complies | |
| 25 g |
Safety and Handling
EINECSNumber : 219-318-8
TSCA : TSCA