missing translation for 'onlineSavingsMsg'
Learn More

o-Cresol, 99%

Catalog No. AC110551000
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
2.5 kg
10 kg
Packaging:
Glass bottle
Plastic drum
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC110551000 100 g Glass bottle
AC110555000 500 g Glass bottle
AC110550010 1 kg Glass bottle
AC110550025 2.5 kg Glass bottle
AC110550100 10 kg Plastic drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. AC110551000 Supplier Thermo Scientific Chemicals Supplier No. 110551000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 95-48-7 | C7H8O | 108.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 95-48-7 , 108-95-2
Molecular Formula C7H8O
Molecular Weight (g/mol) 108.14
MDL Number MFCD00002226
InChI Key QWVGKYWNOKOFNN-UHFFFAOYSA-N
Synonym o-cresol, 2-hydroxytoluene, o-methylphenol, 2-cresol, phenol, 2-methyl, orthocresol, o-cresylic acid, o-oxytoluene, o-toluol, o-hydroxytoluene
PubChem CID 335
ChEBI CHEBI:28054
IUPAC Name 2-methylphenol
SMILES CC1=CC=CC=C1O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 30.0°C to 32.0°C
Color Amber to Colorless
Density 1.0400g/mL
Boiling Point 191.0°C
Flash Point 81°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3C6H4OH
Quantity 100 g
Beilstein 06,349
Merck Index 15,2564
Specific Gravity 1.04
Solubility Information Solubility in water: 20g/L (20°C). Other solubilities: soluble in alcohol,ether and chloroform
Viscosity 4.75 mPa.s (35°C)
Formula Weight 108.14
Percent Purity 99%
Physical Form Liquid or Low Melting Solid
Chemical Name or Material o-Cresol
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Harmful to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN:
EINECSNumber 202-423-8
Recommended Storage Darkens on storage and exposure to light and air

RUO – Research Use Only