missing translation for 'onlineSavingsMsg'
Learn More

O-(2-Azido-4,6-O-benzylidene-2-deoxy-α-D-galactopyranosyl)-N-Fmoc-L-threonine tert-Butyl Ester 97.0+%, TCI America™

Catalog No. a1832100mg
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A1832100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A1832100MG Supplier TCI America Supplier No. A1832100MG

Chemical Identifiers

CAS 195976-07-9
Molecular Formula C36H40N4O9
Molecular Weight (g/mol) 672.735
InChI Key BPJMKNVMUZHIFM-HMAUQUTRSA-N
Synonym O-(2-Azido-4,6-O-benzylidene-2-deoxy-alpha-D-galactopyranosyl)-N-Fmoc-L-threonine tert-Butyl Ester, Fmoc-Thr[GalN3[46Bzd]-alpha]-OtBu
PubChem CID 53384322
IUPAC Name tert-butyl (2S,3R)-3-[[(4aS,6S,7S,8R,8aR)-7-azido-8-hydroxy-2-phenyl-4,4a,6,7,8,8a-hexahydropyrano[3,2-d][1,3]dioxin-6-yl]oxy]-2-(9H-fluoren-9-ylmethoxycarbonylamino)butanoate
SMILES CC(C(C(=O)OC(C)(C)C)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13)OC4C(C(C5C(O4)COC(O5)C6=CC=CC=C6)O)N=[N+]=[N-]

Specifications

Color Yellow
Quantity 100 mg
Formula Weight 672.74
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material O-(2-Azido-4,6-O-benzylidene-2-deoxy-alpha-D-galactopyranosyl)-N-[(9H-fluoren-9-ylmethoxy)carbonyl]-
TSCA No
Recommended Storage Freezer