accessibility menu, dialog, popup

UserName

CAS RN 195976-07-9

IUPAC Name: tert-butyl 3-({7-azido-8-hydroxy-2-phenyl-hexahydro-2H-pyrano[3,2-d][1,3]dioxin-6-yl}oxy)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)butanoate
Synonyms: tert-butyl 3-({7-azido-8-hydroxy-2-phenyl-hexahydro-2H-pyrano[3,2-d][1,3]dioxin-6-yl}oxy)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}butanoate
Molecular Weight (g/mol): 672.74
Molecular Formula: C36H40N4O9
InChi Key: BPJMKNVMUZHIFM-UHFFFAOYNA-N
SMILES: CC(OC1OC2COC(OC2C(O)C1N=[N+]=[N-])C1=CC=CC=C1)C(NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(=O)OC(C)(C)C
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results