missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NuPAGE™ Tris-Acetate Resupply Pack, 3 to 8%

Catalog No. EA0375CRS10
Change view
Click to view available options
Wells:
10-well
12-well
15-well
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Wells
EA0375CRS10 15-well
EA0375ARS10 10-well
EA0375BRS10 12-well
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. EA0375CRS10 Supplier Invitrogen™ Supplier No. EA0375CRS10
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The NuPAGE Tris-Acetate Resupply Pack simplifies your large molecular weight protein separation workflow. This pack, sufficient for 100 gel runs, includes precast mini gels, running buffer, and protein ladders.

The NuPAGE Tris-Acetate Resupply Pack simplifies your large molecular weight protein separation workflow. This pack, sufficient for 100 gel runs, includes precast mini gels, running buffer, and protein ladders. NuPAGE Tris-Acetate gels offer high-resolution protein separation while maintaining protein integrity. Designed for optimal separation and transfer they are ideal for researchers investigating large molecular weight proteins. With a long shelf life this pack is a reliable and convenient solution for your laboratory needs.

The NuPAGE Tris-Acetate Resupply Pack contains:

  • NuPAGE Tris-Acetate gels (10 boxes, 100 gels)
  • NuPAGE Tris-Acetate Running Buffer for 100 gel runs, 20X (4 bottles, Cat. No. LA0041)
  • HiMark Pre-stained Protein Standard for 100 gel runs, 30 to 460 kDa (4 vials, Cat. No. LC5699)

About NuPAGE Tris-Acetate Gels

Invitrogen NuPAGE Tris-Acetate protein gels are precast polyacrylamide gels specifically engineered to provide excellent separation of large molecular weight proteins. These gels do not contain SDS, enabling you to run your proteins in native or denatured form, offering maximum flexibility for diverse protein electrophoresis needs. The unique buffer formulation of these gels maintains a low operating pH during electrophoresis, resulting in superior resolution of proteins when compared to traditional Tris-glycine SDS-PAGE gels.

Not only do NuPAGE Tris-Acetate gels offer high-resolution separation, but they are also designed to ensure optimal transfer of larger molecular weight proteins, making them a preferred choice for researchers performing western blotting. These gels represent a significant advancement over traditional gels, providing both superior performance and flexibility for protein gel electrophoresis and transfer.

Separation

For running Invitrogen protein gels, you have the flexibility to use either the Mini Gel Tank (A25977) or the XCell SureLock Mini-Cell (EI0001). Both systems are designed to provide optimal performance. The Mini Gel Tank's unique side-by-side design offers efficient and convenient electrophoresis for both precast and SureCast handcast mini gels. Alternatively, the XCell SureLock Mini-Cell allows quick, easy, and leak-free running of mini gels, without the need for any clamps. This compatibility gives you the freedom to choose the best tool for your specific lab and experiment requirements. Additionally, the Mini Gel Tank is conveniently available through Invitrogen Precast Gel Welcome Packs along with both sample buffer and reducing agent, providing an all-in-one solution for your electrophoresis needs.

Transfer

For transferring your proteins to a membrane, we recommend using for NuPAGE Transfer Buffer (NP0006) for traditional wet transfer using the Mini Blot Module (B1000). Alternatively, rapid semi-dry transfer can be done using the Invitrogen Power Blotter or rapid dry transfer using the iBlot 3 Gel Transfer Device (IB31001).

Specifications

Content And Storage • NuPAGE Tris-Acetate gels, 10 x 10 gels
• NuPAGE Tris-Acetate Running Buffer (20X), 4 bottles
• HiMark Pre-stained Protein Standard (30 to 460 kDa), 4 vials
Gel Percentage 3 to 8%
Gel Size Mini
Gel Type Tris-Acetate
Separation of Protein
Separation Range 40 to 500 kDa
Separation Type Denaturing, Native
Wells 15-well
Quantity 1 Kit
For Use With (Equipment) Mini Gel Tank

For Research Use Only. Not for use in diagnostic procedures.