missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NuPAGE™ Midi Bis-Tris Resupply Pack, 4 to 12%, WedgeWell™ format

Catalog No. STM5001RS10
Change view
Click to view available options
Wells:
12 + 2-well
20-well
26-well
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Wells
STM5001RS10 12 + 2-well
STM5002RS10 20-well
STM5003RS10 26-well
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. STM5001RS10 Supplier Invitrogen™ Supplier No. STM5001RS10

The NuPAGE Bis-Tris Resupply Pack, Wedgewell format streamlines your protein separation workflow. This pack, sufficient for 100 gel runs, includes precast midi gels, running buffer, and protein ladders. NuPAGE Bis-Tris gels offer high-resolution separation while maintaining protein integrity in a neutral pH environment.

The NuPAGE Bis-Tris Resupply Pack, Wedgewell format streamlines your protein separation workflow. This pack, sufficient for 100 gel runs, includes precast midi gels, running buffer, and protein ladders. NuPAGE Bis-Tris gels offer high-resolution separation while maintaining protein integrity in a neutral pH environment. Designed for optimal separation of a wide range of protein sizes under denaturing conditions, they're ideal for protein sequencing, mass spectrometry, and other techniques where protein integrity is critical. With a long 16-month shelf life and stringent QC process, this pack is a reliable and convenient solution for your laboratory needs.

The NuPAGE Bis-Tris Wedgewell Resupply Pack streamlines your protein separation workflow. This pack, sufficient for 100 gel runs, includes precast midi gels, running buffer, and protein ladders. NuPAGE Bis-Tris gels offer high-resolution separation while maintaining protein integrity in a neutral pH environment. Designed for optimal separation of a wide range of protein sizes under denaturing conditions, they're ideal for protein sequencing, mass spectrometry, and other techniques where protein integrity is critical. With a long 16-month shelf life and stringent QC process, this pack is a reliable and convenient solution for your laboratory needs.

Resupply Pack contents

  • NuPAGE Bis-Tris WedgeWell gels (10 boxes, 100 gels)
  • NuPAGE MES Running Buffer for 100 gel runs, 20X (5 bottles, Cat. No. NP0002)
  • PageRuler Plus Pre-stained Protein Ladder for 100 gel runs, 10 to 250 kDa (26619)

About NuPAGE gels

Invitrogen NuPAGE Bis-Tris protein gels are precast polyacrylamide gels designed for optimal separation of a wide range of proteins under denaturing conditions. Their neutral pH environment is key to minimizing protein modifications, making them ideal for applications where maintaining protein integrity is crucial, such as sequencing and mass spectrometry. These gels offer fast protein separation, delivering results in as little as 35 minutes. Midi-format NuPAGE gels offer increased sample throughput per gel, enhancing efficiency, increasing reproducibility, and decreasing cost per sample. Additionally, their long shelf-life of 12 months at room temperature adds to their practicality and convenience, making them an excellent choice for everyday laboratory use.

Separation

For running Invitrogen protein gels, you have the flexibility to use either the SureLock Tandem Midi Gel Tank (STM1001) or the XCell4 SureLock Midi-Cell (WR0100). Both systems are designed to provide optimal performance. The SureLock Tandem offers rapid and efficient electrophoresis of midi gels in a leak-free system, with minimal buffer usage. The two independent chambers allow for electrophoresis of one or two gels simultaneously, conserving buffer and reducing waste.

Alternatively, the XCell4 SureLock Midi-Cell is a robust and efficient system for midi-gel electrophoresis. Its standout feature is its capacity to simultaneously run up to four midi gels, enabling high-throughput protein analysis with enhanced efficiency and convenience. Additionally, the SureLock Tandem Midi Gel Tank is conveniently available through Invitrogen Precast Gel Welcome packs along with both sample buffer and reducing agent, providing an all-in-one solution for your electrophoresis needs.

Transfer

For transferring your proteins to a membrane, we recommend using for NuPAGE Transfer Buffer (NP0006) for traditional wet transfer using the SureLock Tandem Midi Blot Module (STM2001). Alternatively, rapid semi-dry transfer can be done using the Invitrogen Power Blotter or rapid dry transfer using the iBlot 3 Gel Transfer Device (IB31001).

Convenient to use

NuPAGE Bis-Tris WedgeWell-format gels have innovative wedge-shaped wells that allow up to two times more sample load volume than other 1.0-mm-thick gels. The wedge-shaped wells also have larger openings, making sample loading easier.

Specifications

Content And Storage • NuPAGE Bis-Tris gels, 10 x 10 gels
• NuPAGE MES Running Buffer (20X), 5 bottles
• PageRuler Plus Pre-stained Protein Ladder (10 to 250 kDa)
Gel Percentage 4 to 12%
Gel Size Midi
Gel Type Bis-Tris
Separation of Protein
Separation Range 3.5 to 260 kDa
Separation Type Denaturing
Wells 12 + 2-well
Quantity 1 Kit
For Use With (Equipment) XCell4 SureLock Midi-Cell

For Research Use Only. Not for use in diagnostic procedures.