Learn More
Invitrogen™ NuPAGE™ Bis-Tris IPG-well Welcome Pack
The NuPAGE Bis-Tris IPG-well Welcome Pack provides all of the necessary NuPAGE gels, buffers, and reagents needed to begin using the XCell SureLock Mini-Cell for second-dimension separation in a 2D workflow.
Supplier:  Invitrogen™ NP0330Z
Catalog No. NP0330Z
Only null left
Description
Description
The NuPAGE Bis-Tris IPG-well Welcome Pack provides all of the necessary NuPAGE gels, buffers, and reagents you will need to begin using the XCell SureLock Mini-Cell for your second-dimension separation in a 2D workflow. IPG-well gels are designed to easily accommodate 7-cm IPG strips and a molecular weight marker as well. The XCell SureLock Mini-Cell is a vertical mini-protein gel electrophoresis system compatible with all Invitrogen mini precast gels and Surecast handcast gels.
The NuPAGE Bis-Tris Welcome Pack contains:
• XCell SureLock Mini-Cell (Cat. No. EI0001)
• NuPAGE Bis-Tris mini gels (2 boxes, 20 gels)
• NuPAGE MES SDS Running Buffer, 20X (Cat. No. NP0002)
• NuPAGE LDS Sample Buffer, 4X (Cat. No. NP0007)
• NuPAGE Sample Reducing Agent, 10X (Cat. No. NP0009)
• PageRuler Plus Prestained Protein Ladder, 10 to 250 kDa (Cat. No. 26619)
About the XCell SureLock Mini-Cell
The XCell SureLock Mini-Cell is a vertical mini protein gel electrophoresis system compatible with all Invitrogen mini precast gels and Surecast handcast gels. The Mini-Cell is simple, sturdy, and easy to use. Each XCell SureLock Mini-Cell can accommodate up to two gels per run. Run times may vary depending on buffer conditions and power supplies used.
ZOOM Bis-Tris IPG-Well Precast Gels
ZOOM gels are designed for optimal second dimension separation following isoelectric focusing. They feature an IPG well designed to easily accommodate 7-cm IPG strips and a molecular weight marker as well. Invitrogen IPG well gels are compatible with the XCell SureLock Mini Cell and the Mini Gel Tank.
About NuPAGE Bis-Tris gels
Invitrogen NuPAGE Bis-Tris Protein gels are precast polyacrylamide gels designed to give optimal separation of a wide range of proteins under denaturing conditions. Unlike traditional Tris-glycine gels, NuPAGE Bis-Tris gels have a neutral pH environment that minimizes protein modifications. Use NuPAGE Bis-Tris gels for preparing proteins for sequencing, mass spectrometry, and any other techniques where protein integrity is crucial. Also, use NuPAGE gels for optimal results during day-to-day use.
Specifications
Specifications
4 to 12%
1.0 mm
3.5 to 260 kDa
1 Welcome Pack
Zoom IPG well
XCell SureLock Mini-Cell
Mini
Bis-Tris
Gel Electrophoresis, Isoelectric Focusing (IEF or IPG)
Approved for shipment at Room Temperature or on Wet Ice
NuPAGE
Product Suggestions
Product Suggestions