missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NuPAGE™ Bis-Tris Mini Protein Gels, 4–12%, 1.0–1.5 mm

Catalog No. NP0322BOX
Encompass
Change view
Click to view available options
Wells:
1-well
10-well
12-well
15-well
17-well
2D-well
9-well
IPG-well
Gel Thickness:
1.0 mm
1.5 mm
Quantity:
10 gels (1 box)
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Wells Gel Thickness Quantity
NP0322BOX 12-well 1.0 mm 10 gels (1 box)
NP0321BOX 10-well 1.0 mm 10 gels (1 box)
NP0323BOX 15-well 1.0 mm 10 gels (1 box)
NP0324BOX 1-well 1.0 mm 10 gels (1 box)
NP0326BOX 2D-well 1.0 mm 10 gels (1 box)
NP0327BOX 9-well 1.0 mm 10 gels (1 box)
NP0329BOX 17-well 1.0 mm 10 gels (1 box)
NP0330BOX IPG-well 1.0 mm 10 gels (1 box)
NP0335BOX 10-well 1.5 mm 10 gels (1 box)
NP0336BOX 15-well 1.5 mm 10 gels (1 box)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Catalog No. NP0322BOX Supplier Invitrogen™ Supplier No. NP0322BOX
Only null left

Includes

Catalog numbers for gels refer to a single box. One box contains 10 gels.

Invitrogen NuPAGE Bis-Tris protein gels are precast polyacrylamide gels that provide broad molecular weight protein separation with high resolution and sample integrity.

Invitrogen NuPAGE Bis-Tris protein gels are precast polyacrylamide gels that provide broad molecular weight protein separation with high resolution and sample integrity. These precast gels are ideal for applications where protein integrity is crucial. Unlike traditional Tris-glycine gels, NuPAGE Bis-Tris gels have a neutral pH environment that minimizes protein modifications. Use NuPAGE Bis-Tris gels for preparing proteins for sequencing, mass spectrometry, and any other techniques where protein integrity is important.

Features of NuPAGE Bis-Tris gels:
Better protein integrity—optimized sample preparation process preserves your proteins
Wide ranges of molecular weight separation—select the right gel and running buffer to get the optimal separation of your proteins
Faster run times—get separation of your proteins in as little as 35 minutes
Longer shelf life—NuPAGE Bis-Tris gels can be stored for at least 12 months at room temperature

Choose the right NuPAGE Bis-Tris gel for your protein separation
Obtain optimal separation of your proteins by choosing the right combination of gel and running buffer. NuPAGE Bis-Tris protein gels come in four polyacrylamide concentrations: 8%, 10%, 12%, and a 4–12% gradient. Gels come in two sizes: mini (8 cm x 8 cm) or midi (8.7 cm x 13.3 cm) and either 1.0 mm (mini and midi gels) or 1.5 mm (mini gel format only) in thickness. NuPAGE Bis-Tris gels also come in multiple well formats.

NuPAGE Bis-Tris gels are formulated for denaturing gel electrophoresis applications. For optimal sample preparation, use the NuPAGE LDS Sample Buffer (NP0007) and NuPAGE Sample Reducing Agent (NP0004). Use NuPAGE Antioxidant (NP0005) in the running buffer to maintain the reduced state of the proteins during the run and to allow maximum band sharpness. The gels can be run using NuPAGE MES SDS Running Buffer (NP0002) to better resolve small proteins or NuPAGE MOPS SDS Running Buffer (NP000102) to resolve medium- to large-size proteins.

For transfer of proteins to a membrane, we recommend using NuPAGE Transfer Buffer (NP00061) for traditional wet transfer using the Mini Blot Module (B1000) or XCell II Blot Module (EI9051). Alternatively, rapid semi-dry transfer can be done using the Invitrogen Power Blotter or rapid dry transfer using the iBlot 2 Gel Transfer Device (IB21001).

Specifications

Gel Percentage 4 to 12%
Gel Size Mini
Gel Thickness 1.0 mm
Gel Type Bis-Tris
Recommended Applications Denaturing
Sample Loading Volume up to 20 μL
Separation Range 3.5 to 260 kDa
Separation Type Denaturing
Wells 12-well
Mode of Separation Molecular Weight
Quantity 10 gels (1 box)
Shipping Condition Room Temperature
Storage Requirements Store at 4–25°C. Do not freeze.
Length (Metric) 8 cm
Width (Metric) 8 cm
Product Line NuPAGE
For Use With (Equipment) Mini Gel Tank
Show More Show Less