Learn More
Invitrogen™ NuPAGE™ Bis-Tris Mini Protein Gels, 10%, 1.0–1.5 mm
Invitrogen NuPAGE Bis-Tris protein gels are precast polyacrylamide gels that provide broad molecular weight protein separation with high resolution and sample integrity. These precast gels are ideal for applications where protein integrity is crucial.
Supplier: Invitrogen™ NP0302BOX
Includes: One box of 10 gels
Description
Invitrogen NuPAGE Bis-Tris protein gels are precast polyacrylamide gels that provide broad molecular weight protein separation with high resolution and sample integrity. These precast gels are ideal for applications where protein integrity is crucial. Unlike traditional Tris-glycine gels, NuPAGE Bis-Tris gels have a neutral pH environment that minimizes protein modifications. Use NuPAGE Bis-Tris gels for preparing proteins for sequencing, mass spectrometry, and any other techniques where protein integrity is important.
Features of NuPAGE Bis-Tris gels:
• Better protein integrity—optimized sample preparation process preserves your proteins
• Wide ranges of molecular weight separation—select the right gel and running buffer to get the optimal separation of your proteins
• Faster run times—get separation of your proteins in as little as 35 minutes
• Longer shelf life—NuPAGE Bis-Tris gels can be stored for at least 12 months at room temperature
Choose the right NuPAGE Bis-Tris gel for your protein separation
Obtain optimal separation of your proteins by choosing the right combination of gel and running buffer. NuPAGE Bis-Tris protein gels come in four polyacrylamide concentrations: 8%, 10%, 12%, and a 4–12% gradient. Gels come in two sizes: mini (8 cm x 8 cm) or midi (8.7 cm x 13.3 cm) and either 1.0 mm (mini and midi gels) or 1.5 mm (mini gel format only) in thickness. NuPAGE Bis-Tris gels also come in multiple well formats.
NuPAGE Bis-Tris gels are formulated for denaturing gel electrophoresis applications. For optimal sample preparation, use the NuPAGE LDS Sample Buffer (NP0007) and NuPAGE Sample Reducing Agent (NP0004). Use NuPAGE Antioxidant (NP0005) in the running buffer to maintain the reduced state of the proteins during the run and to allow maximum band sharpness. The gels can be run using NuPAGE MES SDS Running Buffer (NP0002) to better resolve small proteins or NuPAGE MOPS SDS Running Buffer (NP000102) to resolve medium- to large-size proteins.
For transfer of proteins to a membrane, we recommend using NuPAGE Transfer Buffer (NP00061) for traditional wet transfer using the Mini Blot Module (B1000) or XCell II Blot Module (EI9051). Alternatively, rapid semi-dry transfer can be done using the Invitrogen Power Blotter or rapid dry transfer using the iBlot 2 Gel Transfer Device (IB210010).
Order Info
Shipping Condition: Room Temperature
Chemical Identifiers
| 10% | |
| Bis-Tris | |
| Denaturing | |
| Room Temperature | |
| 8 cm | |
| NuPAGE |
| Mini | |
| 3.5 to 260 kDa | |
| Molecular Weight | |
| Store at 4–25°C. Do not freeze. | |
| 8 cm |
Specifications
| 10% | |
| 1.0 mm | |
| Denaturing | |
| 3.5 to 260 kDa | |
| 12-well | |
| 10 gels (1 box) | |
| Store at 4–25°C. Do not freeze. | |
| 8 cm | |
| XCell SureLock Mini-Cell |
| Mini | |
| Bis-Tris | |
| up to 20 μL | |
| Denaturing | |
| Molecular Weight | |
| Room Temperature | |
| 8 cm | |
| NuPAGE |
Safety and Handling
ShelfLife : 12 Months