missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nunc™ Square BioAssay Dishes

Catalog No. 12565224
Change view
Click to view available options
Height (Metric):
20 mm
25 mm
Surface Treatment:
Non-treated
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Height (Metric) Surface Treatment
12565224 25 mm Non-treated
12565743 20 mm Non-treated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 12565224 Supplier Thermo Scientific™ Supplier No. 240835
Only null left

Screen large numbers of colonies for cloning experiments, culturing bacteria, fungi and agar diffusion assays using Thermo Scientific™ Nunc™ Squre BioAssay dishes.

Available in cell culture-treated and non-treated surfaces, the versatile Thermo Scientific™ Nunc™ Square BioAssay dishes are useful for screening large numbers of colonies, culturing bacteria and fungi, or providing a contained environment for other culture vessels.

  • Use as a moisture chamber for incubation on Nunc MicroWell™ Plates
  • Lined with filter paper it is ideal as an incubation chamber for in situ hybridization
  • Optically clear and compatible with automated robotic instrumentation

Untreated Dishes

  • Ideal for screening libraries or isolating clones
  • Useful for screening large numbers of colonies in cloning experiments
  • Can be used for culturing of bacteria and fungi as well as a moisture chamber for incubation of Nunc MicroWell Plates
  • Suitable in agar diffusion assay
  • Lined with wet filter paper, it is also useful as an incubation chamber for in situ hybridization
  • Large growth area
  • Designed for genomic screen and colony picking

Specifications

Bottom Shape Flat
Capacity (Metric) 225 mL
Culture Area 500 sq. cm
Shape Square
Sterility Sterile
Surface Treatment Non-treated
Type Multi-Well Plate
Volume (Metric) Working 225 mL
No. per Case 16
No. per Pack 4
Height (Metric) 25 mm
Height (English) 25 in.
Show More Show Less

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.