missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Nunc™ MicroWell™ 96-Well, Nunclon Delta-Treated, Flat-Bottom Microplate
Color: Clear; Sterility: Sterile; Lid: With Lid
$1,636.18 - $4,520.68
Specifications
| Barcode | Custom Barcoding Available |
|---|---|
| Lid | With Lid |
| Array | 8 x 12 |
| Product Line | Nunc |
| Color | Clear |
| Catalog Number | Mfr. No. | No. of Wells | No. per Pack | No. per Case | Price | Quantity & Availability | |||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | No. of Wells | No. per Pack | No. per Case | Price | Quantity & Availability | |||
|
1256566
|
Thermo Scientific™
167008 |
96 | 1 Microplate | 50 Microplates |
Case of 50 Each for $1,636.18
|
|
|||
|
12565383
|
Thermo Scientific™
161093 |
96 | 10 Microplates | 160 Microplates |
Case of 160 Each for $4,520.68
|
|
|||
Description
Thermo Scientific™ Nunc MicroWell 96-Well Microplates are optimized for robotics, automated readers and liquid handling systems. 96-well microplates are used for antibiotic screens, cell-based assays and screening compounds. The flat bottom shape is ideal for microscopic and optical measurements. Our plates are high quality for consistent and reliable results.
- Clear polystyrene has excellent optical properties making it ideal for optical measurements
- Cell Culture Treated: The Nunclon™ Delta cell culture treated surface is a hydrophilic surface that promotes cell attachment and growth
- Plates are optimized for robotics, plate readers and liquid handlers
- Customized bar coding is available
Specifications
| Custom Barcoding Available | |
| 8 x 12 | |
| Clear | |
| Polystyrene | |
| 96 | |
| Nunclon Delta |
| With Lid | |
| Nunc | |
| Microplate | |
| 400 μL | |
| Sterile | |
| Flat Bottom (F-well) |