Learn More
Thermo Scientific™ Nunc™ MicroWell™ 96-Well, Nunclon Delta-Treated, Flat-Bottom Microplate
Color: Clear; Sterility: Sterile; Lid: With Lid
Supplier:  Thermo Scientific™ 161093
 View more versions of this product
Catalog No. 12565383
Only null left
Description
Description
Thermo Scientific™ Nunc MicroWell 96-Well Microplates are optimized for robotics, automated readers and liquid handling systems. 96-well microplates are used for antibiotic screens, cell-based assays and screening compounds. The flat bottom shape is ideal for microscopic and optical measurements. Our plates are high quality for consistent and reliable results.
  • Clear polystyrene has excellent optical properties making it ideal for optical measurements
  • Untreated polystyrene is hydrophobic and is suited for growth of suspension cultures that can proliferate and grow without attachment
  • Untreated surfaces are ideal for adding coatings in your lab
  • Cell Culture Treated: The Nunclon™ Delta cell culture treated surface is a hydrophilic surface that promotes cell attachment and growth
  • Plates are optimized for robotics, plate readers and liquid handlers
  • Customized bar coding is available
Specifications
Chemical Identifiers
Custom Barcoding Available
8 x 12
Clear
Polystyrene
96
Nunclon Delta
With Lid
Nunc
Microplate
400 μL
Sterile
Flat Bottom (F-well)
Specifications
Custom Barcoding Available
With Lid
8 x 12
Nunc
Microplate
Polystyrene
96
Nunclon Delta
160 Microplates
160/Cs.
All microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004)
96F well plate,Nunclon Delta, clear, with lid, Sterile (Pack of 10, Case of 160)
10 Pk., 160 Cases
Clear
High Flange
400 μL
Sterile
Flat Bottom (F-well)
10 Microplates
Product Suggestions
Product Suggestions