Learn More
Thermo Scientific™ Nunc™ Lab-Tek™ Petri Dishes
Description
Culture fungi, bacteria and other microorganisms with Thermo Scientific™ Nunc™ Lab-Tek™ Petri Dishes.
- Available with a molded grid
- Performs well in automatic dispensers
- Contact dish useful for sampling in hospital environments as well as in food science and the pharmaceutical industry
- Compatible with automated systems
Specifications
Specifications
| Description | Vented, without stack ring |
| Bottom Shape | Flat |
| Culture Area | 79 sq. cm |
| Shape | Square |
| Type | Petri Dish |
| Well Count | 1 |
| Vent | Yes |