missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ Nukol™ Capillary GC Column, df 1 μm

Useful for the analyses of volatile acidic compounds.

Supplier:  MilliporeSigma™ Supelco™ SGMA25357

 View more versions of this product

Catalog No. 111003272


Only null left
Explore available promotions

Description

Description

Application: The incorporation of acid functional groups into the phase lends an acidic character to this column, useful for analyses of volatile acidic compounds. Difficult to analyze carboxylic acids (free fatty acids) can be analyzed with excellent peak shape and minimal adsorption. USP Code: This column meets USP G25 and G35 requirements. Phase: Bonded. Acid-modified poly(ethylene glycol). Temp. Limits:. 60 C to 200 C (isothermal) or 220 C (programmed)
Specifications

Specifications

0.53 mm
Nukol
1 μm
Acid-modified poly(ethylene glycol)
GC Accessories & Adsorbents
Gas Chromatography (GC)
30 m
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

Nukol is a trademark of Sigma-Aldrich Co. LLC