missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nuclear Fast Red, Spectrum™ Chemical
$337.82 - $33744.57
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
18-612-059
|
Spectrum Chemical Mfg Cor
NU1051GM |
1 g | Amber Glass Bottle |
Each for $337.82
|
|
||||
|
18-612-062
|
Spectrum Chemical Mfg Cor
NU1055GM |
5 g | Amber Glass Bottle |
Each for $934.32
|
|
||||
|
18-612-061
|
Spectrum Chemical Mfg Cor
NU10525GM |
25 g | Amber Glass Bottle |
Each for $2,343.14
|
|
||||
|
18-612-063
|
Spectrum Chemical Mfg Cor
NU105500GMBL |
500 g | Amber Glass Bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
18-612-060
|
Spectrum Chemical Mfg Cor
NU1051KGBL |
1 kg | Amber Glass Bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
| 6409-77-4 |
Specifications
| 6409-77-4 | |
| Reagent | |
| Dark Red | |
| Nuclear Fast Red |
| 1 | |
| Amber Glass Bottle | |
| 1 g |
CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.