accessibility menu, dialog, popup

UserName

CAS RN 6409-77-4

IUPAC Name: sodium 4-amino-1,3-dihydroxy-9,10-dioxo-9,10-dihydroanthracene-2-sulfonate
Synonyms: sodium 4-amino-1,3-dihydroxy-9,10-dioxoanthracene-2-sulfonate
Molecular Weight (g/mol): 357.27
Molecular Formula: C14H8NNaO7S
InChi Key: IFSXZLJQEKGQAF-UHFFFAOYSA-M
SMILES: [Na+].NC1=C(O)C(=C(O)C2=C1C(=O)C1=CC=CC=C1C2=O)S([O-])(=O)=O
Quantity 
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
Grade 
  • (1)
  • (2)
  • (5)
Color 
  • (5)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (7)
  • (5)
  • (1)
  • (5)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (5)
  • (5)
  • (1)
  • (1)