missing translation for 'onlineSavingsMsg'
Learn More

Nuclear Fast Red, Spectrum™ Chemical

Catalog No. 18612059
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
500 g
1 kg
Packaging:
Amber Glass Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18612059 1 g Amber Glass Bottle
18612062 5 g Amber Glass Bottle
18612061 25 g Amber Glass Bottle
18612063 500 g Amber Glass Bottle
18612060 1 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18612059 Supplier Spectrum Chemical Mfg Cor Supplier No. NU1051GM
Only null left

Chemical Identifiers

CAS 6409-77-4

Specifications

CAS Max % 1
Grade Reagent
Packaging Amber Glass Bottle
Color Dark Red
Quantity 1 g
Chemical Name or Material Nuclear Fast Red

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.