missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ NS 8593 Hydrochloride

Catalog No. 459750
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
459750 50 mg
459710 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 459750 Supplier R&D Systems™ Supplier No. 4597/50

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A Tocris product, part of R&D Systems. Selective negative modulator of KCa2 (SK) channels; inhibits SK currents

NS 8593 hydrochloride is a selective KCa2 (SK) channel negative modulator; inhibits SK channel currents (Kd values are 0.42, 0.6 and 0.73 μM for SK1, SK2 and SK3 respectively at 0.5 μM Ca2+). Exhibits selectivity for SK channels over KCa1.1 (BK), KCa3.1 (IK), Kv, Nav and Cav channels. Inhibits afterhyperpolarization in hippocampal slices.

Specifications

Chemical Name or Material NS 8593 hydrochloride
CAS 875755-24-1
Synonym N-[(1R)-1,2,3,4-Tetrahydro-1-naphthalenyl]-1H-benzimidazol-2-amine hydrochloride
Quantity 50 mg
Solubility Soluble to 100mM in DMSO and to 20mM in ethanol
Product Line Tocris
Molecular Formula C17H17N3·HCl
Formula Weight 299.8
Physical Form Solid
Product Type Bioactive Small Molecule
Purity >98%
Show More Show Less

Research Use Only. Not For Human or Veterinary Use