missing translation for 'onlineSavingsMsg'
Learn More

Innovatek NRSII™ Transwab™ Environmental Swabs

Innovatek NRSII™ Transwab™ Environmental Swabs accurately recover residual bacteria on food contact surfaces

$2,320.75

Specifications

Media Type Neutralizers
Certifications/Compliance ISO 18593
Closure Color High Visibility Blue
Color Clear
For Use With (Application) Sampling Previously Cleaned Surfaces, Food Safety
View More Specs

Products 3
Catalog Number Mfr. No. Volume (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Volume (Metric) Price Quantity & Availability  
361041858
View Documents Promotion Details
Innovatek
4080MW786
1 mL
Case of 500 Each for $2,320.75
Only null left
 
361041856
View Documents Promotion Details
Innovatek
4080MW784
5 mL
Case of 500 Each for $2,320.75
Only null left
 
361041857
View Documents Promotion Details
Innovatek
4080MW785
10 mL
Case of 500 Each for $2,320.75
Only null left
 
Description

Description

  • Swab is preimmersed, ready for use with a self standing tube
  • High visibility blue cap and swab is easy to find and retrieve if accidentally dropped
  • Conforms to ISO 18593 and other national and international standards
  • Neutralisers allow accurate recovery of residual bacteria on food contact surfaces
  • Precise fill for accurate assessment of contamination levels
  • Reduced packaging & storage space
  • Ambient storage and transport
Specifications

Specifications

Neutralizers
High Visibility Blue
Sampling Previously Cleaned Surfaces, Food Safety
Ambient Conditions
ISO 18593
Clear
50/Pk.
Videos
SDS
Documents

Documents