Learn More
Innovatek NRSII™ Transwab™ Environmental Swabs
Innovatek NRSII™ Transwab™ Environmental Swabs accurately recover residual bacteria on food contact surfaces
Supplier: Innovatek 4080MW784
Description
- Swab is preimmersed, ready for use with a self standing tube
- High visibility blue cap and swab is easy to find and retrieve if accidentally dropped
- Conforms to ISO 18593 and other national and international standards
- Neutralisers allow accurate recovery of residual bacteria on food contact surfaces
- Precise fill for accurate assessment of contamination levels
- Reduced packaging & storage space
- Ambient storage and transport
Chemical Identifiers
| Neutralizers | |
| High Visibility Blue | |
| Sampling Previously Cleaned Surfaces, Food Safety | |
| Ambient Conditions |
| ISO 18593 | |
| Clear | |
| 50/Pk. |
Specifications
| Neutralizers | |
| High Visibility Blue | |
| Sampling Previously Cleaned Surfaces, Food Safety | |
| Ambient Conditions |
| ISO 18593 | |
| Clear | |
| 50/Pk. | |
| 5 mL |