missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ NPS 2390
Description
Group I mGlu antagonist; displays noncompetitive antagonist activity at both mGlu1 and mGlu5 receptors. Thought to act on a site separate from the glutamate binding pocket.
Specifications
Specifications
| Quantity | 10 mg |
| Formula Weight | 307.39 |
| Purity | >99% |
| Molecular Weight (g/mol) | 307.397 |
| Chemical Name or Material | NPS 2390 |
Safety and Handling
| Recommended Storage | Store at Room Temperature |