missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ NP-40 Surfact-Amps™ Detergent Solution

Catalog No. p-6014004
Change view
Click to view available options
Quantity:
50 mL
500 mL
6 x 10 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI85124 50 mL
PI28324 6 x 10 mL
PI85125 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. PI85124 Supplier Thermo Scientific™ Supplier No. 85124
Only null left

Thermo Scientific™ Surfact-Amps NP-40 Detergent Solution is highly-purified NP-40 detergent stabilized and supplied as a 10%solution.

Thermo Scientific Surfact-Amps NP-40 Detergent Solution is highly-purified NP-40 detergent stabilized as a 10% solution in your choice of either 10 mL glass ampules or two sizes of HDPE-plastic bottles.

Features of Surfact-Amps NP-40 Detergent Solution:

  • NP-40—a nonionic detergent for use in various protein methods
  • Accurate—precise 10% detergent solutions in ultrapure water
  • Easy-to-use—solutions are simple to dispense and dilute for use
  • Exceptionally pure—less than 1.0μeq/mL peroxides and carbonyls
  • Stable—packaged under inert nitrogen gas in HDPE bottles

This is an easy-to-use 10% (w/v) solution of purified NP-40 detergent for use in routine and high-demand protein research methods and molecular biology techniques. NP-40 is a nonionic polyoxyethylene surfactant that is most frequently used as a component of cell lysis buffers or other solutions intended to extract and solubilize proteins. Surfact-Amps Detergent Solutions (10% w/v) provide unsurpassed purity, quality and stability. Unlike neat detergents, which are extremely viscous, Surfact-Amps 10% Solutions are easy to pipette and accurately dispense. The surfactant solutions are carefully prepared and packaged under nitrogen in glass ampules or non-leaching HDPE bottles, ensuring their stability and eliminating the accumulation of peroxides and degradation products.

Properties of NP-40 Detergent:

  • Molecular Weight: 617 g
  • Detergent Class: Nonionic
  • Aggregation Number: 149
  • Micelle Molecular Weight: 90,000 g
  • Critical Micelle Concentration (CMC): 0.29 mM (0.0179%, w/v)
  • Cloud Point: 80°C
  • Dialyzable: No

Specifications for NP-40 Surfact-Amps Detergent Solution:

  • Visual: Clear, colorless solution, free of particulate matter.
  • Concentration: 10.0±1.0%
  • Oxidants: ≤1.0μeq/mL
  • Carbonyls: ≤1.0μeq/mL
  • Suspended Solids: Residue present must not exceed Residue Reference.

Related Products

  • Surfact-Amps™ Detergent Sampler (Cat. No. 28340)

Specifications

Product Type Detergent
Content And Storage Store between 20°C and 25°C
Form Liquid
Product Line Surfact-Amps
Reagent Type Detergent Solution
Quantity 50 mL
Format HDPE Bottle

For Research Use Only. Not for use in diagnostic procedures.