missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Novex™ Tris-Glycine ZOOM™ Mini Protein Gel, 4 to 20%, 1.0 mm, IPG-well
Separation Range: 20 to 200 kd; Separation Type: Denaturing; For Use With: ZOOM IPGRunner Mini
Supplier: Invitrogen™ EC60261BOX
Includes: One box of 10 gels
Description
- Used to separate proteins based upon their molecular weight after isoelectric focusing (IEF)
- 2D gels are generally designed to accept IEF tube gels, sample lanes sliced from vertical or horizontal IEF slab gels, or immobilized pH gradient (IPG) strips
- Gels are usually run under denaturing conditions using SDS in the gel running buffers
- The IPG well accommodates the full length of a 7cm IPG strip
- In addition to the IPG well, ZOOM Gels have a separate well designed for use with a molecular weight standard
- Standard 10cm x 10cm cassette format
Sample Loading Volume: 7cm strips; IPG Well
2D Gel Electrophoresis, Protein Gel Electrophoresis, Proteins, Expression, Isolation and Analysis
Order Info
Shipping Condition: Wet Ice
Specifications
| One box contains 10 gels. Store in refrigerator (2–8°C). Do not freeze. Shelf life is 8 weeks. | |
| Mini | |
| Tris-Glycine | |
| Protein | |
| Denaturing | |
| Molecular Weight | |
| Wet Ice | |
| IPG Well | |
| 8 cm | |
| 8 cm | |
| Gradient Page | |
| ZOOM |
| 4 to 20% | |
| 1.0 mm | |
| 7 cm strips | |
| 20 to 200 kDa | |
| 1-well | |
| 10 gels (1 box) | |
| 1.0 mm | |
| 8 cm | |
| 8 cm | |
| Novex, ZOOM | |
| ZOOM IPGRunner Mini |
For Research Use Only. Not for use in diagnostic procedures.