Learn More
Invitrogen™ Novex™ WedgeWell™ Tris-Glycine Welcome Pack, 10 to 20%
The Novex Tris-Glycine Welcome Pack provides all of the necessary Novex Tris-glycine gels, buffers, and reagents needed to begin using the Mini Gel Tank.
$748.00 - $750.00
Specifications
| For Use With (Equipment) | Mini Gel Tank |
|---|---|
| Gel Percentage | 10 to 20% |
| Gel Size | Mini |
| GelThickness | 1.0 mm |
| Gel Type | Tris-Glycine |
| Catalog Number | Mfr. No. | Wells | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Wells | Price | Quantity | |||||
XP1020A
|
Invitrogen™
XP1020A |
10-well |
Pack for $750.00
|
|
|||||
XP1020B
|
Invitrogen™
XP1020B |
12-well |
Pack for $750.00
|
|
|||||
XP1020C
|
Invitrogen™
XP1020C |
15-well |
Pack for $748.00
|
|
|||||
Description
The Novex Tris-Glycine Welcome Pack provides all of the necessary Novex Tris-glycine gels, buffers, and reagents you need to begin using the Mini Gel Tank. The Mini Gel Tank is compatible with all Novex and NuPAGE mini gels. Each Mini Gel Tank can accommodate up to two gels per run. The unique tank design enables convenient, side-by-side gel loading and enhanced viewing during use. Run times may vary depending on buffer conditions and power supplies being used.
The Novex Tris-Glycine Welcome Pack contains:
• Mini Gel Tank (A25977)
• Novex Tris-Glycine Mini Gels, WedgeWell format (2 boxes, 20 gels)
• Novex Tris-Glycine SDS Running Buffer, 10X (LC2675)
• Novex Tris-Glycine SDS Sample Buffer, 2X (LC2676)
• NuPAGE Sample Reducing Agent, 10X (NP0004)
• PageRuler Plus Prestained Protein Ladder, 10 to 250 kDa (26619)
About the Mini Gel Tank
The Mini Gel Tank is compatible with all Novex and NuPAGE mini gels. Each Mini Gel Tank can accommodate up to two gels per run. The unique tank design enables convenient, side-by-side gel loading and enhanced viewing during use. Run times may vary depending on buffer conditions and power supplies being used.
About Novex Tris-Glycine gels, WedgeWell format
Novex Tris-Glycine mini gels are polyacrylamide gels based on traditional Laemmli protein electrophoresis, allowing the use of Laemmli sample and running buffers. They enable consistent performance and separation of a wide range of proteins into well-resolved bands. Novex Tris-Glycine gels, WedgeWell format, have innovative wedge-shaped wells that allow up to two times more sample load volume than other 1.0 mm-thick gels. The wedge-shaped wells also have larger openings, making sample loading easier.
For transferring your proteins to a membrane, we recommend using for Novex Tris-Glycine Native Running Buffer (LC2672) or Novex Tris-Glycine Transfer Buffer (LC3675) for traditional wet transfer using the Mini Blot Module (B1000). Alternatively, rapid semi-dry transfer can be done using the Invitrogen Power Blotter or rapid dry transfer using the iBlot 2 Gel Transfer Device (IB21001).
Specifications
| Mini Gel Tank | |
| Mini | |
| Tris-Glycine | |
| Novex, WedgeWell | |
| Wet Ice |
| 10 to 20% | |
| 1.0 mm | |
| 6 to 200 kDa | |
| 1 Welcome Pk. kit |