missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Novex™ Tris-Glycine SDS Running Buffer (10X) DFS Item

Formulated for separation of proteins, in their denatured state, on Novex Tris-Glycine gels

$73.00 - $259.00

Specifications

Chemical Name or Material Running Buffer
Concentration 10X
Product Line Novex
View More Specs

Includes: Novex Tris-Glycine SDS Running Buffer (10X), 500mL

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
LC2675 DFS Item
View Documents Promotion Details
Invitrogen™
LC2675
500 mL
Each for $73.00
Only null left
 
LC26754 DFS Item
View Documents
Invitrogen™
LC26754
4 x 1 L
Pack of 4 Each for $248.00
Only null left
 
LC26755 DFS Item
View Documents
Invitrogen™
LC26755
5 L
Each for $259.00
Only null left
 
Description

Description

Tris-Glycine gels provide reproducible separation of a wide range of proteins into well-resolved bands, depending upon the gel percentage used.

  • Pre-mixed buffers are a convenient way to ensure high-quality, consistent electrophoresis results
  • Made with high-purity reagents and are strictly quality controlled

1D Gel Electrophoresis, Protein Gel Electrophoresis, Proteins, Expression, Isolation and Analysis

Order Info

Shipping Condition: Room Temperature

Specifications

Specifications

Running Buffer
10X
Novex
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

For Research Use Only. Not for use in diagnostic procedures.